Learn More
Kova™ POC Urinalysis Dropper Controls
Description
- Designed for use in POC sites and smaller labs
- Control values assigned for the most commonly used dipsticks and instruments
- After opening, control is stabile for 30 days at room temp or until expiration when stored in refrigerator
- Barcoded labels (lot/expiration)
- Ready-to-use liquid controls in 2 levels (Normal/Abnormal)
- 22 month shelf life from date of manufacture
Specifications
Specifications
| Certifications/Compliance | CLSI and CLIA, ANSI Z400.5-2004 |
| Color | Yellow, green or amber |
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | pH, Protein, Glucose, Ketones, Bilirubin, Hemoglobin, Nitrite, Urobilinogen, Specific Gravity, Leukocytes, Microalbumin, Creatinine, Ascorbic Acid |
| For Use With (Equipment) | Urinalysis reagent strips |
| Intended Use | Ready-to-use, liquid products intended for use as a control for qualitative and semi-quantitative procedures used in routine urinalysis |
| Quantity | 10 x 5 mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.