Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POC1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559217
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559217 Supplier Invitrogen™ Supplier No. PA559217
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLLYTLHGHQ GPATTVAFSR TGEYFASGGS DEQVMVWKSN FDIVDHGEVT KVPRPPATLA SSMGNLPEVD FPVPPGRGRS VESVQSQPQE PVSVPQTLTS TLEH Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

POC1 proteins contain an N-terminal WD40 domain and a C-terminal coiled coil domain and are part of centrosomes. They play an important role in basal body and cilia formation. This gene encodes one of the two POC1 proteins found in humans. Mutations in this gene result in short stature, onychodysplasia, facial dysmorphism, and hypotrichosis (SOFT) syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POC1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POC1A
Gene Accession No. Q8NBT0
Gene Alias 2510040D07Rik; hypothetical protein LOC534489; Pix2; POC1 centriolar protein A; POC1 centriolar protein homolog A; POC1 centriolar protein homolog A (Chlamydomonas); Poc1a; Proteome of centriole protein 1A; RGD1565004; SOFT; WD repeat domain 51A; WD repeat-containing protein 51A; Wdr51a
Gene Symbols POC1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POC1A. Recombinant protein control fragment (Product #RP-98202).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25886
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.