Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POC1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110190 Supplier Invitrogen™ Supplier No. PA5110190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LFSLSQHINW VRCAKFSPDG RLIVSASDDK TVKLWDKSSR ECVHSYCEHG GFVTYVDFHP SGTCIAAAGM DNTVKVWDVR THRLLQHYQL HSAAVNGLSF HPSGNYLITA SSDSTLKILD LMEGRLLYTL HGHQGPATTV AFSRTGEYFA SGGSDEQVMV WKSNFDIVDH GEVTKVPRPP ATLASSMGNL PEVDFPVPPG RGRSVESVQS QPQEPVSVPQ TLTSTLEHIV GQLDVLTQTV SILEQRLTLT EDKLKQCLEN QQLIMQRATP.

POC1 proteins contain an N-terminal WD40 domain and a C-terminal coiled coil domain and are part of centrosomes. They play an important role in basal body and cilia formation. This gene encodes one of the two POC1 proteins found in humans. Mutations in this gene result in short stature, onychodysplasia, facial dysmorphism, and hypotrichosis (SOFT) syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POC1A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POC1A
Gene Accession No. Q8JZX3, Q8NBT0
Gene Alias 2510040D07Rik; hypothetical protein LOC534489; Pix2; POC1 centriolar protein A; POC1 centriolar protein homolog A; POC1 centriolar protein homolog A (Chlamydomonas); Poc1a; Proteome of centriole protein 1A; RGD1565004; SOFT; WD repeat domain 51A; WD repeat-containing protein 51A; Wdr51a
Gene Symbols POC1A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 138-407 of human POC1A (NP_056241.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25886, 501048, 70235
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.