Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POC1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577223
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
POC1 proteins contain an N-terminal WD40 domain and a C-terminal coiled coil domain and are part of centrosomes. They play an important role in basal body and cilia formation. This gene encodes one of the two POC1 proteins found in humans. Mutations in this gene result in short stature, onychodysplasia, facial dysmorphism, and hypotrichosis (SOFT) syndrome.
Specifications
POC1A | |
Polyclonal | |
Unconjugated | |
POC1A | |
2510040D07Rik; hypothetical protein LOC534489; Pix2; POC1 centriolar protein A; POC1 centriolar protein homolog A; POC1 centriolar protein homolog A (Chlamydomonas); Poc1a; Proteome of centriole protein 1A; RGD1565004; SOFT; WD repeat domain 51A; WD repeat-containing protein 51A; Wdr51a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25886, 501048, 70235 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8JZX3, Q8NBT0 | |
POC1A | |
Recombinant full length Human POC1A. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction