Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POC1A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549028
Description
Predicted to react with zebrafish based on sequence homology.
Plays an important role in centriole assembly and/or stability and ciliogenesis. Involved in early steps of centriole duplication, as well as in the later steps of centriole length control. Acts in concert with POC1B to ensure centriole integrity and proper mitotic spindle formation.
Specifications
POC1A | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8NBT0, Q8JZX3 | |
POC1A | |
KLH conjugated synthetic peptide between 34-61 amino acids from the N-terminal region of human POC1A. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
POC1A | |
POC1 centriolar protein homolog A, Pix2, Proteome of centriole protein 1A, WD repeat-containing protein 51A, POC1A, WDR51A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25886, 70235 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction