Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POC1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113291
Description
POC1B Polyclonal Antibody for Western Blot, IHC (P), ELISA
Required for ciliogenesis.
Specifications
| POC1B | |
| Polyclonal | |
| Unconjugated | |
| POC1B | |
| 4933430F16Rik; CORD20; Pix1; POC1 centriolar protein B; POC1 centriolar protein homolog B; POC1 centriolar protein homolog B (Chlamydomonas); POC1B; Proteome of centriole protein 1B; TUWD12; WD repeat domain 51B; WD repeat-containing protein 51B; WDR51B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 282809, 382406 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BHD1, Q8TC44 | |
| POC1B | |
| Recombinant Human POC1 centriolar protein homolog B protein (299-478AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction