Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POC1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524495
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524495 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524495 Supplier Invitrogen™ Supplier No. PA524495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POC1B Polyclonal Antibody for IHC (P), Western Blot

Required for ciliogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POC1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene POC1B
Gene Accession No. Q8TC44
Gene Alias 4933430F16Rik; CORD20; Pix1; POC1 centriolar protein B; POC1 centriolar protein homolog B; POC1 centriolar protein homolog B (Chlamydomonas); POC1B; Proteome of centriole protein 1B; TUWD12; WD repeat domain 51B; WD repeat-containing protein 51B; WDR51B
Gene Symbols POC1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 321-350 amino acids from the C-terminal region of human WDR51B.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282809
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.