Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POC5 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178741B
Description
POC5 Polyclonal antibody specifically detects POC5 in Human samples. It is validated for Western BlotSpecifications
POC5 | |
Polyclonal | |
Western Blot | |
centrosomal protein POC5, FLJ35779, POC5 centriolar protein homolog (Chlamydomonas) | |
The immunogen for this product maps to a region between residue 525 and 575 of human Protein of Centriole 5 using the numbering given in entry NP_001092741.1 (GeneID 134359). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
134359 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction