Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583375
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583375 Supplier Invitrogen™ Supplier No. PA583375
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KWEEYEELLH YAIVTPNIEP CASQSSHPKG ELVPDVRIST IHDILHSQGN NSEVRETAIE VGKGCDFHIS SHSKTDESSP VLSPRKPSHP VMD Highest antigen sequence indentity to the following orthologs: Rat - 51%, Mouse - 56%.

Essential for the assembly of the distal half of centrioles, required for centriole elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POC5
Applications Western Blot, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POC5
Gene Accession No. Q8NA72
Gene Alias 1200014M14Rik; AV144706; C5orf37; Centrosomal protein POC5; hPOC5; Poc5; POC5 centriolar protein; POC5 centriolar protein homolog; POC5 centriolar protein homolog (Chlamydomonas); protein of centriole 5; proteome of centriole 5; RGD1310597
Gene Symbols POC5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POC5. Recombinant protein control fragment (Product #RP-104673).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134359
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.