Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POD1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988709
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8709 Supplier Bioss Supplier No. BS8688R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The basic helix-loop-helix transcription factor TCF21/Pod1 (also called capsulin or epicardin) is involved in kidney, lung and spleen organogenesis. It is also essential for normal development of the testes and ovaries. TCF21/Pod1 is involved in the transcriptional repression of steroidogenic factor 1 (Sf1/Nr5a1/Ad4BP), an orphan nuclear receptor that regulates the expression of multiple genes (including Scc) that mediate sexual differentiation.

Specifications

Antigen POD1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TCF21
Gene Accession No. O43680
Gene Alias BHLHA23; BOS_9830; Capsulin; Class A basic helix-loop-helix protein 23; epc; Epicardin; hypothetical protein LOC558148; LOC100346722; Msf-1; MyoRa2; Pod1; Pod-1; Podocyte-expressed 1; tcf21; TCF-21; Transcription factor 21; zgc:123313
Gene Symbols TCF21
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TCF-21/POD1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6943
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.