Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PODN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531625 Supplier Invitrogen™ Supplier No. PA531625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Jurkat. Predicted reactivity: Human (99%), Mouse (95%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Non-collagenous proteins of the small leucine-rich repeat (SLR) protein family are considered important components of the extracellular matrix. The extracellular matrix plays an integral role in the pivotal processes of development, tissue repair and metastasis by regulating cell proliferation, differentiation, adhesion and migration. Podocan, also known as PCAN, SLRR5A or PODN, is a 613 amino acid secreted protein belonging to the small leucine-rich proteoglycan (SLRP) family and the class V subfamily. Expressed in kidney, heart, liver, pancreas and vascular smooth muscle cells, Podocan exists as three alternatively spliced isoforms containing twenty LRR (leucine-rich repeats), a unique N-terminal cysteine-rich cluster pattern and a highly acidic C-terminal domain. Podocan is considered a glycoprotein containing N-linked oligosaccharides that may be involved in growth regulation in cardiovascular tissues. Encoded by a gene located on human chromosome 1, Podocan interacts type I collagen (COL1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PODN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Podn
Gene Accession No. Q7Z5L7
Gene Alias 9430070G18; Pcan; Podn; Podocan; podocan proteoglycan; PRO332; SLRR5A; UNQ293; UNQ293/PRO332
Gene Symbols Podn
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PODN. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127435
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.