Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podocalyxin/GCTM-1 Monoclonal Antibody (ZY165), Abbomax

Catalog No. 502204392
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4392 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4392 Supplier Abbomax Inc Supplier No. 605850
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Breast carcinoma Cellular location: Apical cell membrane.

Podocalyxin/GCTM-1 Monoclonal antibody specifically detects Podocalyxin/GCTM-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Podocalyxin/GCTM-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ZY165
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene PODXL
Gene Accession No. O00592
Gene Alias AW121214; GCTM-2 antigen; Gp200; Ly102; MGC138240; PC; PCLP; PCLP antibody; Pclp1; PCLP-1; PCLP1 antibody; PODLX2; podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; podolyxin; Podxl
Host Species Mouse
Immunogen A partial recombinant protein of human GCTM-1
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5420
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.