Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podocalyxin (GP135) Mouse anti-Canine, Unconjugated, Clone: 3F2:D8, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS132725UL
Description
Anti-Podocalyxin (GP135), clone 3F2:D8, Cat. No. MABS1327, is a mouse monoclonal antibody that detects Podocalyxin (GP135) and has been tested for use in Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Radioimmunoassay, and Western Blotting.
Specifications
Podocalyxin (GP135) | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
PODXL;PCLP;PCLP1 | |
Intact Madin-Darby canine kidney (MDCK) cells. | |
Protein G purified | |
RUO | |
Primary | |
Canine | |
Antibodies | |
IgG1 κ |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoradiometric Assay, Western Blot | |
3F2:D8 | |
Unconjugated | |
Podocalyxin-like protein 1;GP1345;PC;PC-like protein 1;PCLP-1;cPCLP1 | |
Mouse | |
180 kda obseerved; 59.91 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
25 μL | |
Signaling | |
Clone 3F2:D8 detects Podocalyxin in Madin-Darby canine kidney (MDCK) cells. | |
Stable for 1 year at 2-8°C from date of receipt. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction