Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podocalyxin Like Antibody (2A4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$610.00
Specifications
Antigen | Podocalyxin Like |
---|---|
Clone | 2A4 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry : 5 - 10 ul/million cells in 0.1ml, Immunohistochemistry-Paraffin : 1:50 - 1:100, Immunofluorescence : 1:25 - 1:50, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247750A
![]() |
Novus Biologicals
NBP2477500.1MG |
0.1 mg |
Each for $610.00
|
|
|||||
NBP247750B
![]() |
Novus Biologicals
NBP2477500.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Podocalyxin Like Monoclonal specifically detects Podocalyxin Like in Human, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence, CyTOF-ready.Specifications
Podocalyxin Like | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cellular Markers, Embryonic Stem Cell Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
PBS with No Preservative | |
5420 | |
A recombinant protein fragment containing the intracellular, transmembrane, and part of the extracellular domain of human podocalyxin. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
2A4 | |
Flow Cytometry : 5 - 10 ul/million cells in 0.1ml, Immunohistochemistry-Paraffin : 1:50 - 1:100, Immunofluorescence : 1:25 - 1:50, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Rabbit | |
Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1 | |
PODXL | |
IgM | |
Protein A or G purified | |
Podocalyxin is a member of the CD34 transmembrane sialomucin family. It is over-expressed on the podocyte foot projections and plays essential roles in kidney development and homeostasis, blood filtration and urine formation. It is also expressed on vascular endothelia, hematopoietic progenitors and a subset of neurons. Overexpression of podocalyxin may be linked to more aggressive tumor behavior. Podocalyxin antibody can identify podocytes in the urine (podocyturia) that may indicate glomerular disease, pre-eclampsia, and other kidney pathology. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title