Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podocalyxin Like Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11041503AF700
Description
Podocalyxin Like Polyclonal specifically detects Podocalyxin Like in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).Specifications
Podocalyxin Like | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
O00592 | |
PODXL | |
A synthetic peptide within an internal region (residues 100-200) of the human Podocalyxin protein. [Swiss-Prot: O00592] This immunogen should be far enough away from any glycosylation site. | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Embryonic Stem Cell Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
5420 | |
Store at 4C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction