Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podocalyxin Like Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$206.50 - $499.50
Specifications
Antigen | Podocalyxin Like |
---|---|
Concentration | 0.90 mg/mL |
Dilution | Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:1000 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11041503T
![]() |
Novus Biologicals
NB11041503SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB11041503
![]() |
Novus Biologicals
NB11041503 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Podocalyxin Like Polyclonal specifically detects Podocalyxin Like in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).Specifications
Podocalyxin Like | |
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:1000 | |
Polyclonal | |
Rabbit | |
Cancer, Cell Biology, Cellular Markers, Embryonic Stem Cell Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
Tris-Glycine, 0.15 M NaCl with 0.05% Sodium Azide | |
Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1 | |
PODXL | |
IgG | |
Affinity Purified |
0.90 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
Unconjugated | |
RUO | |
Human, Mouse | |
O00592 | |
5420 | |
A synthetic peptide within an internal region (residues 100-200) of the human Podocalyxin protein. [Swiss-Prot: O00592] This immunogen should be far enough away from any glycosylation site. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title