Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podocalyxin Like Antibody (OTI2C6), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273511R
Description
Podocalyxin Like Monoclonal specifically detects Podocalyxin Like in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Podocalyxin Like | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PODXL | |
| Recombinant protein fragment corresponding to amino acids 32-461 of human Podocalyxin Like (NP_005388) produced in HEK293 cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2C6 | |
| Western Blot, Immunohistochemistry | |
| Gp200MGC138240, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, podocalyxin-like, Podocalyxin-like protein 1 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cell Biology, Cellular Markers, Embryonic Stem Cell Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| 5420.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction