Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Podocin Recombinant Rabbit Monoclonal Antibody (JB51-33)
GREENER_CHOICE

Catalog No. PIMA534674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534674 Supplier Invitrogen™ Supplier No. MA534674
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Rat kidney tissue, human kidney tissue, mouse kidney tissue, 293T.

The pathological conditions of SRN1 correlate well with mutations at the NPHS2 gene, where expression of a protein known as Podocin occurs. Abnormal or inefficient signaling through Podocin protein-dependent networks contributes to the development of podocyte dysfunction and proteinuria. The human NPHS2 gene maps to chromosome 1q25-q31 and encodes a 383 amino acid protein. Podocin is an integral membrane protein that appears to fold into a hairpin-like structure with intracellular amino- and carboxy-termini. Transmembrane and cytoplasmic portions of Podocin share homology to the corresponding regions of the stomatin family proteins. Expression of high-order oligomers of Podocin in glomerular podocytes may reflect a scaffolding function that influences proper function of the glomerular filtration barrier, which is necessary for renal stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Podocin
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JB51-33
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Nphs2
Gene Accession No. Q91X05, Q9NP85
Gene Alias AI790225; nephrosis 2 homolog, podocin; nephrosis 2 idiopathic steroid-resistant (podocin); nephrosis 2, idiopathic, steroid-resistant; nephrosis 2, idiopathic, steroid-resistant (podocin); nephrosis 2, podocin; NPHS2; NPHS2 podocin; NPHS2 stomatin family member, podocin; NPHS2, podocin; PDCN; podocin; SRN1
Gene Symbols Nphs2
Host Species Rabbit
Immunogen Synthetic peptide within Human NPHS2 aa 334-383.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170484, 7827
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.