Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podophyllum Resin, USP, 40-50%, Spectrum™ Chemical
SDP

Catalog No. 18605253
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle

PO105, 9000-55-9

Spectrum™ Chemical Podophyllum Resin, USP is used as a medication to treat genital warts and plantar warts. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9000-55-9
Molecular Formula C88H92O34
Molecular Weight (g/mol) 1693.67
InChI Key KOUZYZCESNTXJO-UHFFFAOYNA-N
IUPAC Name bis(16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one); bis(8-hydroxy-7-(hydroxymethyl)-5-(3,4,5-trimethoxyphenyl)-2H,5H,6H,7H,8H-naphtho[2,3-d][1,3]dioxole-6-carboxylic acid)
SMILES COC1=CC(=CC(OC)=C1OC)C1C2C(COC2=O)C(O)C2=CC3=C(OCO3)C=C12.COC1=CC(=CC(OC)=C1OC)C1C2C(COC2=O)C(O)C2=CC3=C(OCO3)C=C12.COC1=CC(=CC(OC)=C1OC)C1C(C(CO)C(O)C2=CC3=C(OCO3)C=C12)C(O)=O.COC1=CC(=CC(OC)=C1OC)C1C(C(CO)C(O)C2=CC3=C(OCO3)C=C12)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.015
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 40 to 50%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.