Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Antibody (070), Alexa Fluor™ 594

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290499AF594
Description
Podoplanin Monoclonal specifically detects Podoplanin in Mouse samples. It is validated for ELISA.Specifications
Podoplanin | |
Monoclonal | |
Alexa Fluor 594 | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Podoplanin (NP_034459.2; Met1-Lys133). | |
0.1 mL | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
10630 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
70 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction