Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Antibody (8.1.1), Alexa Fluor™ 594, Novus Biologicals™

Golden Syrian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NB6001015AF594
Description
Podoplanin Monoclonal specifically detects Podoplanin in Mouse, Human (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Podoplanin | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PDPN | |
Murine thymic stromal cell lines | |
0.1 mL | |
Primary | |
Mouse, Human | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
8.1.1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
Golden Syrian Hamster | |
Protein G purified | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
10630.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction