Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Antibody (LF3/B7/D5B27) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$281.00 - $572.50
Specifications
| Antigen | Podoplanin |
|---|---|
| Clone | LF3/B7/D5B27, B7, D5B3 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1-5 ug/ml, Immunohistochemistry 6-30 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 6-30 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1096423SS
![]() |
Novus Biologicals
NB11096423SS |
0.025 mg |
Each for $281.00
|
|
|||||
NB11096423
![]() |
Novus Biologicals
NB11096423 |
0.1 mg |
Each for $572.50
|
|
|||||
Description
Podoplanin Monoclonal specifically detects Podoplanin in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Podoplanin | |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Rat | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| PDPN | |
| IgG1 | |
| Protein G purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LF3/B7/D5B27, B7, D5B3 | |
| Western Blot 1-5 ug/ml, Immunohistochemistry 6-30 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 6-30 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q64294 | |
| 10630 | |
| Membrane protein fraction of isolated rat glomeruli | |
| Primary | |
| Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title