Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Antibody (NZ-1.2), DyLight 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP203952UV
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
Podoplanin | |
Monoclonal | |
DyLight 350 | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
NZ-1.2 | |
Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
PDPN | |
A synthetic peptide corresponding to amino acids 38-70 from the platelet-aggregation-stimulating (PLAG) domain of human podoplanin was used as immunogen. | |
0.1 mL | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
10630 | |
Store at 4C in the dark. | |
IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction