Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Antibody (PMab-1), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP311971R
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
Podoplanin | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
IgG κ |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
PMab-1 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
This antibody was raised by immunising rats with 14-residue synthetic peptide mpp3851, which corresponds to amino acids 38-51 (GDGMVPPGIEDKIT) of the platelet-aggregation-stimulating (PLAG) domain of mouse podoplanin. Spleen cells were then harvested, and fused to P3U1 cells to generate stable hybridomas. | |
0.1 mL | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
10630 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction