Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Antibody (RTD4E10), mFluor Violet 610 SE, Novus Biologicals™

Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP236058MFV610
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse, Human (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunoprecipitationSpecifications
Podoplanin | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Hamster | |
Protein G purified | |
RUO | |
Primary | |
Mouse, Human (Negative) | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
RTD4E10 | |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
Murine thymic stromal cell line | |
0.1 mL | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
10630 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction