Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin, Mouse anti-Human, Clone: LpMab-13, Millipore Sigma™

Catalog No. MABT85525UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT85525UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT85525UG Supplier MilliporeSigma Supplier No. MABT85525UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Podoplanin, clone LpMab-13, Cat. No. MABT855, is a mouse monoclonal antibody that detects human podoplanin and has been tested for use in ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), and Western Blotting.

Podoplanin (UniProt: Q86YL7; also known as Aggrus, Glycoprotein 36, Gp36, PA2.26 antigen, T1-alpha, T1A) is encoded by the PDPN (also known as GP36) gene (Gene ID: 10630) in human. Podoplanin serves as the endogenous ligand of C-type lectin-like receptor-2 (CLEC-2) and is highly expressed in various tumors and in some normal cells, such as lymphatic endothelial cells and podocytes. It may be involved in cell migration and/or actin cytoskeleton organization. Podoplanin is localized to actin-rich microvilli and plasma membrane projections, such as filopodia, lamellipodia, and ruffles. Six isoforms of podoplanin have been described that are produced by alternative splicing. It is synthesized with a signal peptide of 22 amino acids, which is cleaved to produce the mature form. Podoplanin contains an extracellular domain (aa 23-131), a helical domain (aa 132-152) and a cytoplasmic region (aa 153-162). Podoplanin possesses three tandem repeat of platelet aggregation-stimulating (PLAG) domains in its N-terminus. Among the PLAG domains, sialylated O-glycan on Thr52 of PLAG3 is essential for the binding to C-type lectin-like receptor-2 (CLEC-2). Interaction of human podoplanin with CLEC-2 mainly involves Glu47 and Asp48 in the platelet PLAG3. Clone LpMab-13 binds to PLAG2/3 domains and recognizes endogenous hPDPN of cancer cells, including glioblastoma, oral cancer, lung cancer, and malignant mesothelioma, and normal cells such as lymphatic endothelial cells and podocytes of kidney. The interaction between LpMab-13 and hPDPN is independent of its glycosylation. The minimum epitope of LpMab-13 is identified to be Ala42-Asp49 of hPDPN. ((Ref.: Ogasawara, S., et al. (2016). Monoclon. Antib. immunodiag. Immunother. 35(3); 155-163).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Podoplanin
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone LpMab-13
Conjugate Unconjugated
Dilution ELISA Analysis: A representative lot detected Podoplanin in ELISA application (Ogasawara, S., et. al. (2016). Monoclon Antib Immunodiagn Immunother . 35(3):155-62). Flow Cytometry Analysis: A representative lot detected Podoplanin in Flow Cytometry application (Ogasawara, S., et. al. (2016). Monoclon Antib Immunodiagn Immunother . 35(3):155-62). Immunohistochemistry Analysis: A representative lot detected Podoplanin in Immunohistochemistry application (Ogasawara, S., et. al. (2016). Monoclon Antib Immunodiagn Immunother . 35(3):155-62). Western Blotting Analysis: A representative lot detected Podoplanin in Western Blotting application (Ogasawara, S., et. al. (2016). Monoclon Antib Immunodiagn Immunother . 35(3):155-62).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Aggrus;Glycoprotein 36;Gp36;PA2.26 antigen;T1-alpha;T1A
Gene Symbols PDPN;GP36;PSEC0003;PSEC0025
Host Species Mouse
Immunogen LN229 glioma cells expressing human Podoplanin.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone LpMab-13 detects Podoplanin in human cells. The minimal epitope is shown to be Ala42-Asp49.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.