Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin Mouse, anti-Human, R718, Clone: LPMAB-23, BD OptiBuild™

Catalog No. BDB751982 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751982 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751982 Supplier BD Biosciences Supplier No. 751982
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The LpMab-23 monoclonal antibody specifically recognizes a cancer-specific form of human podoplanin. Podoplanin is a 38-44 kDa type I transmembrane glycoprotein that is encoded by PDPN. This heavily glycosylated mucin type protein is named for its expression on kidney glomerular epithelial cells known as podocytes. Although the podoplanin protein is expressed on a wide variety of cell types in normal tissues (including placenta, lung, skeletal muscle and brain), LpMab-23 recognizes an altered glycosylation pattern that occurs on oral cancer cells and it shows minimal reactivity with the surrounding non-cancerous tissue. In contrast, the LpMab-17 monoclonal antibody recognizes a non-glycosylated epitope of podoplanin on normal and cancer cells. Podoplanin induces platelet aggregation via its three platelet aggregation-stimulating (PLAG) domains| it binds C-type lectin domain family 1 member B (Clec1B, also known as CLEC2)| and it is involved in actin cytoskeleton organization and in cellular adhesion and migration. It also plays roles in organogenesis, vascular development, inflammatory diseases, tumorigenesis, and cancer cell motility and metastasis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Podoplanin
Applications Flow Cytometry
Classification Monoclonal
Clone LPMAB-23
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.