Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin, Mouse anti-Rat, Clone: PMab-2, Millipore Sigma™

Catalog No. MABT85025UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT85025UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT85025UG Supplier MilliporeSigma Supplier No. MABT85025UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Podoplanin, clone PMab-2, Cat. No. MABT850, is a mouse monoclonal that detects rat podoplanin and has been tested for use in ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), and Western Blotting.

Podoplanin (UniProt: Q64294; also known as E11 antigen epitope, Glycoprotein 38, Gp38, OTS-8, RTI140, T1-alpha, T1A, Type I cell 40 kDa protein) is encoded by the Pdpn (also known as Gp38, Ots8) gene (Gene ID: 54320) in rat. Podoplanin serves as the endogenous ligand of C-type lectin-like receptor-2 (CLEC-2) and is highly expressed in various tumors and in some normal cells. It may be involved in cell migration and/or actin cytoskeleton organization. Podoplanin is localized to actin-rich microvilli and plasma membrane projections, such as filopodia, lamellipodia, and ruffles. It is synthesized with a signal peptide of 22 amino acids, which is cleaved to produce the mature form. Podoplanin contains an extracellular domain (aa 23-135), a helical domain (aa 136-156) and a cytoplasmic region (aa 157-166). Podoplanin is extensively O-glycosylated and contains sialic acid residues. O-glycosylation is believed to be essential for its platelet aggregation activity. Clone PMab2 exhibits high affinity for podoplanin (Kd = 0.59 nM). (Ref.: Oki, H et al. (2015). Monoclon. Antib. Immunodiagn. Immunother. 34(6); 396-403).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Podoplanin
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone PMab-2
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Podoplanin in rat spleen tissue. Flow Cytometry Analysis: A representative lot detected Podoplanin in Flow Cytometry applications (Oki, H., et. al. (2015).   Monoclon Antib Immunodiagn Immunother . 34(6):396-403). Western Blotting Analysis: A representative lot detected Podoplanin in Western Blotting applications (Oki, H., et. al. (2015).   Monoclon Antib Immunodiagn Immunother . 34(6):396-403). Immunohistochemistry Analysis: A representative lot detected Podoplanin in Immunohistochemistry applications (Oki, H., et. al. (2015).   Monoclon Antib Immunodiagn Immunother . 34(6):396-403). ELISA Analysis: A representative lot detected Podoplanin in ELISA applications (Oki, H., et. al. (2015).   Monoclon Antib Immunodiagn Immunother . 34(6):396-403).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias E11 antigen epitope;Glycoprotein 38;Gp38;OTS-8;RTI140;T1-alpha;T1A;Type I cell 40 kDa protein
Gene Symbols Pdpn;Gp38;Ots8
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal region of rat Podopalnin.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone PMab-2 detects podoplanin in rat spleen and kidney. It targets an epitope within 14 amino acids from the N-terminal region.
Target Species Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.