Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin Rabbit anti-Mouse, Clone: 66, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$366.00 - $572.50

Specifications

Antigen Podoplanin
Clone 66
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB020588
SDP
View Documents
Novus Biologicals
NBP28955250UL
50 μg
Each for $366.00
Only null left
Add to Cart
 
NB020587
SDP
View Documents
Novus Biologicals
NBP289552100UL
100 μg
Each for $572.50
Only null left
Add to Cart
 
Description

Description

Podoplanin Monoclonal specifically detects Podoplanin in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Podoplanin
Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000
Monoclonal
Purified
RUO
Mouse
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha
Recombinant Mouse Podoplanin Protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
66
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Core ESC Like Genes, Signal Transduction, Stem Cell Markers
0.2 um filtered solution in PBS
10630
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.