Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Podoplanin Rabbit anti-Mouse, Clone: PMab-1, Novus Biologicals™

Rabbit Monoclonal Antibody
$280.00 - $596.50
Specifications
Antigen | Podoplanin |
---|---|
Clone | PMab-1 |
Concentration | 1 mg/mL |
Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Block/Neutralize |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Neutralization |
Description
Podoplanin Monoclonal antibody specifically detects Podoplanin in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, NeutralizationSpecifications
Podoplanin | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Neutralization | |
Unconjugated | |
RUO | |
Mouse | |
36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
This antibody was raised by immunising rats with 14-residue synthetic peptide mpp3851, which corresponds to amino acids 38-51 (GDGMVPPGIEDKIT) of the platelet-aggregation-stimulating (PLAG) domain of mouse podoplanin. Spleen cells were then harvested, and fused to P3U1 cells to generate stable hybridomas. | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
PMab-1 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Block/Neutralize | |
Monoclonal | |
Rabbit | |
Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
PBS | |
10630 | |
IgG κ | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title