Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PODXL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582001 Supplier Invitrogen™ Supplier No. PA582001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPETMSSSPT AASTTHRYPK TPSPTVAHES NWAKCEDLET QTQSEKQLVL NLTGNTLCAG GASDEKLISL ICRAVKATFN PAQDKCGIRL ASVPGSQTVV VKEITIHTKL PAKDVYERLK DKWDELKEAG VSDMKLGD Highest antigen sequence indentity to the following orthologs: Rat - 41%, Mouse - 43%.

This gene encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PODXL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PODXL
Gene Accession No. O00592
Gene Alias AW121214; GCTM-2 antigen; Gp200; Ly102; MGC138240; PC; PCLP; PCLP antibody; Pclp1; PCLP-1; PCLP1 antibody; PODLX2; podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; podolyxin; Podxl
Gene Symbols PODXL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PODXL. Recombinant protein control fragment (Product #RP-101012).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5420
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.