Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PODXL Polyclonal Antibody
PODXL Polyclonal Antibody
Supplier: Thermo Scientific PA583778
Description
PODXL Polyclonal Antibody for Western Blot, IHC (P)

Specifications
PODXL | |
Polyclonal | |
Unconjugated | |
PODXL | |
AW121214; GCTM-2 antigen; Gp200; Ly102; MGC138240; PC; PCLP; PCLP antibody; Pclp1; PCLP-1; PCLP1 antibody; PODLX2; podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; podolyxin; Podxl | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5420 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00592 | |
PODXL | |
Recombinant protein corresponding to Human PODXL. Recombinant protein control fragment (Product #RP-101013). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction