Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PODXL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542988 Supplier Invitrogen™ Supplier No. PA542988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EEEELNDSSL DLGPTADYVF PDLTEKAGSI EDTSQAQELP NLPSPLPKMN Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Podolyxin is a CD34 family member expressed by podocytes, vascular endothelium, mesothelium, and a subset of hematopoietic progenitors. It is the major sialoprotein in the glycocalyx of glomerular podocytes. The physiologic role of podolyxin in leukemic blasts is unknown. However, it is thought of as a candidate prostate cancer tumor aggressiveness gene. Podolyxin appears to complement CD34 as a useful cell surface marker for hemangioblasts, the common precursors of hematopoietic and endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PODXL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PODXL2
Gene Accession No. Q9NZ53
Gene Alias D130074J02Rik; EG; endoglycan; LOW QUALITY PROTEIN: podocalyxin-like protein 2; PODLX2; podocalyxin like 2; podocalyxin-like 2; podocalyxin-like protein 2; Podxl2; RGD1305815; sialomucin protein; UNQ1861/PRO3742
Gene Symbols PODXL2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PODXL2 (aa 101-150).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50512
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.