Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POF1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570750 Supplier Invitrogen™ Supplier No. PA570750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%.

Plays a key role in the organization of epithelial monolayers by regulating the actin cytoskeleton. May be involved in ovary development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POF1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene POF1B
Gene Accession No. Q8WVV4
Gene Alias 2310066B14Rik; POF; POF1B; POF1B actin binding protein; POF1B, actin binding protein; POF2B; premature ovarian failure 1B; premature ovarian failure protein 1B; premature ovarian failure, 1B; protein POF1B; RGD1560798
Gene Symbols POF1B
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human POF1B.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79983
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.