Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POFUT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597014 Supplier Invitrogen™ Supplier No. PA597014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SWDPAGYLLY CPCMGRFGNQ ADHFLGSLAF AKLLNRTLAV PPWIEYQHHK PPFTNLHVSY QKYFKLEPLQ AYHRVISLED FMEKLAPTHW PPEKRVAYCF EVAAQRSPDK KTCPMKEGNP FGPFWDQFHV SFNKSELFTG ISFSASYREQ WSQRRENHSC VTLLFPR; Positive Samples: HepG2, HL-60, Mouse heart, Mouse liver; Cellular Location: Endoplasmic reticulum.

POFUT1, an endoplasmic reticulum-residing member of the glycosyltransferase O-Fuc family, adds O-fucose through an O-glycosidic linkage to conserved serine or threonines in epidermal growth factor-like repeats of several cell surface and secreted proteins. Unlike its homolog POFUT2, POFUT1 can also catalyze the transfer of fucose to thrombospondin type 1 repeats. Many of the substrates of POFUT1 are involved in ligand-induced receptor signaling. One such protein is Notch; mouse ES cells lacking POFUT have normal levels of Notch receptors at the cell surface, but these receptors do not bind Notch ligands or exhibit Notch signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POFUT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene POFUT1
Gene Accession No. Q6EV70, Q91ZW2, Q9H488
Gene Alias DDD2; FUT12; GDP-fucose protein O-fucosyltransferase 1; KIAA0180; mKIAA0180; o-fucosyltransferase protein; O-Fuc-T; OFUCT1; O-FucT-1; O-FucT-1a; O-FucT-1b; O-FUT; peptide-O-fucosyltransferase 1; Pofut1; pofut1 {ECO:0000312; protein O-fucosyltransferase 1; protein O-fucosyltransferase 1 isoform 1 precursor; protein O-fucosyltransferase 1a; protein O-fucosyltransferase 1b; protein-O-fucosyltransferase 1; RGD:1303001}; zgc:110472
Gene Symbols POFUT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 28-194 of human POFUT1 (NP_7584361).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140484, 23509, 311551
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.