Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POFUT2 Antibody (OTI3A12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273512F
Description
POFUT2 Monoclonal specifically detects POFUT2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
POFUT2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
POFUT2 | |
Full length recombinant protein of human POFUT2 (NP_598368) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Flow Cytometry | |
OTI3A12 | |
Flow Cytometry | |
C21orf80, chromosome 21 open reading frame 80, FUT13EC 2.4.1.221, GDP-fucose protein O-fucosyltransferase 2, KIAA0958, O-FucT-2, Peptide-O-fucosyltransferase 2, protein O-fucosyltransferase 2 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
23275.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction