Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POFUT2 Antibody (OTI3A12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245363
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry
POFUT2 Monoclonal specifically detects POFUT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
POFUT2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry 1:100 | |
C21orf80, chromosome 21 open reading frame 80, FUT13EC 2.4.1.221, GDP-fucose protein O-fucosyltransferase 2, KIAA0958, O-FucT-2, Peptide-O-fucosyltransferase 2, protein O-fucosyltransferase 2 | |
Mouse | |
49.8 kDa | |
0.1 mL | |
Lipid and Metabolism | |
23275 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Flow Cytometry | |
OTI3A12 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
POFUT2 | |
Full length human recombinant protein of human POFUT2(NP_598368) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction