Learn More
POFUT2 Mouse anti-Human, Clone: OTI5H1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505004

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Fucose is typically found as a terminal modification of branched chain glycoconjugates, but it also exists in direct O-linkage to serine or threonine residues within cystine knot motifs in epidermal growth factor-like repeats or thrombospondin type-1 repeats. POFUT2 is an O-fucosyltransferase that use THBS type-1 repeats as substrates.Specifications
POFUT2 | |
Monoclonal | |
Unconjugated | |
POFUT2 | |
C21orf80, FUT13 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23275 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI5H1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y2G5 | |
POFUT2 | |
Full length human recombit protein of human POFUT2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.