Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POFUT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110192
Description
Immunogen sequence: MVFARHLREV GDEFRSRHLN STDDADRIPF QEDWMKMKVK LGSALGGPYL GVHLRRKDFI WGHRQDVPSL EGAVRKIRSL MKTHRLDKVF VATDAVRKEY EELKKLLPEM VRFEPTWEEL ELYKDGGVAI I.
Fucose is typically found as a terminal modification of branched chain glycoconjugates, but it also exists in direct O-linkage to serine or threonine residues within cystine knot motifs in epidermal growth factor-like repeats or thrombospondin type-1 repeats. POFUT2 is an O-fucosyltransferase that use THBS type-1 repeats as substrates.
Specifications
| POFUT2 | |
| Polyclonal | |
| Unconjugated | |
| POFUT2 | |
| 2310011G23Rik; AI256847; BC003494; C21orf80; FUT13; GDP-fucose protein O-fucosyltransferase 2; KIAA0958; O-FucT-2; peptide-O-fucosyltransferase 2; POFUT2; protein O-fucosyltransferase 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23275, 309686, 80294 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.53 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8VHI3, Q9Y2G5 | |
| POFUT2 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction