Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POGK Antibody (OTI6D5), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273513APC
Description
POGK Monoclonal antibody specifically detects POGK in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
POGK | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey | |
Purified |
Western Blot, Immunohistochemistry | |
OTI6D5 | |
Western Blot, Immunohistochemistry | |
BASS2, KIAA1513, KIAA15131, KRAB box domain containing 2, KRBOX2, LST003, pogo transposable element with KRAB domain | |
Full length human recombinant protein of human POGK(NP_060012) produced in HEK293T cell. | |
0.1 mL | |
Apoptosis | |
57645 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction