Learn More
Invitrogen™ POGK Monoclonal Antibody (OTI3E2)

Description
POGK Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | POGK |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3E2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | POGK |
| Gene Accession No. | Q9P215 |
| Gene Alias | BASS2; EGK_01919; KIAA1513; KRAB box domain containing 2; KRBOX2; LST003; POGK; pogo transposable element derived with KRAB domain; pogo transposable element with KRAB domain; SLTP003 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.