Learn More
POGK Mouse anti-Human, Clone: OTI3D2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505670

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The exact function of the protein encoded by this gene is not known. However, this gene product contains a KRAB domain (which is involved in protein-protein interactions) at the N-terminus, and a transposase domain at the C-terminus, suggesting that it may belong to the family of DNA-mediated transposons in human.Specifications
POGK | |
Monoclonal | |
Unconjugated | |
POGK | |
BASS2, KRBOX2, LST003 | |
Mouse | |
Affinity Chromatography | |
RUO | |
57645 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3D2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9P215 | |
POGK | |
Full length human recombit protein of human POGK produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.