Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POGK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556804 Supplier Invitrogen™ Supplier No. PA556804
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLQNAHAMRR AFRGPKNGRF ALVDQRVAEY VRYMQAKGDP ITREAMQLKA LEIAQEMNIP EKGFKASLGW CRRMMRRYDL SLRHKVPVPQ HLPEDLTEKL VTYQRSVLAL RRAHDYEVAQ MGNADETPIC LEVPSRVTV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

This gene encodes a 134 kDa protein named strumpellin that is predicted to have multiple transmembrane domains and a spectrin-repeat-containing domain. This ubiquitously expressed gene has its highest expression in skeletal muscle. The protein is named for Strumpell disease; a form of hereditary spastic paraplegia (HSP). Spastic paraplegias are a diverse group of disorders in which the autosomal dominant forms are characterized by progressive, lower extremity spasticity caused by axonal degeneration in the terminal portions of the longest descending and ascending corticospinal tracts. More than 30 loci (SPG1-33) have been implicated in hereditary spastic paraplegia diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POGK
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POGK
Gene Accession No. Q9P215
Gene Alias BASS2; EGK_01919; KIAA1513; KRAB box domain containing 2; KRBOX2; LST003; POGK; pogo transposable element derived with KRAB domain; pogo transposable element with KRAB domain; SLTP003
Gene Symbols POGK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POGK. Recombinant protein control fragment (Product #RP-89161).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57645
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.