Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POGLUT1 Monoclonal Antibody (737512)
GREENER_CHOICE

Catalog No. PIMA524350
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524350 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524350 Supplier Thermo Scientific Supplier No. MA524350
Add to Cart
Add to Cart

POGLUT1 Monoclonal Antibody (737512)

POGLUT1 Monoclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen POGLUT1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 737512
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene POGLUT1
Gene Accession No. Q8NBL1
Gene Alias 9630046K23Rik; C3orf9; CAP10-like 46 kDa protein; CAP10-like protein; CLP46; hCLP46; hRumi; KDELC family like 1; KDELCL1; KTEL (Lys-Tyr-Glu-Leu) containing 1; KTEL motif-containing protein 1; KTELC1; MDS010; MDSRP; Myelodysplastic syndromes relative protein; O-glucosyltransferase Rumi homolog; Poglut1; Protein O-glucosyltransferase 1; protein O-xylosyltransferase; Protein O-xylosyltransferase POGLUT1; RGD1306248; Rumi; UNQ490/PRO1006; Wing-shaped neural plate protein; Wsnp; x 010 protein
Gene Symbols POGLUT1
Host Species Mouse
Immunogen CHO-derived recombinant human Protein O-Glucosyltransferase 1/POGLUT Arg24-Leu388.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56983
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.