Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POGLUT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113132 Supplier Invitrogen™ Supplier No. PA5113132
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POGLUT1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

KTELC1 also termed as CLP46 is a glucosyltransferase, which implicates in Protein modification and glycosylation. KTELC1 encodes a 392 amino acids protein and its molecular weight is 46 kDa, but is exists glycosylation modification. KTELC1 is localized in endoplasmic reticulum lumen and abundantly expressed in liver, brain, heart, skeletal muscle, spleen, kidney, placenta, lung and peripheral blood leukocyte. KTELC1 may regulation in cell proliferation and Notch signal pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POGLUT1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene POGLUT1
Gene Accession No. Q8NBL1
Gene Alias 9630046K23Rik; C3orf9; CAP10-like 46 kDa protein; CAP10-like protein; CLP46; hCLP46; hRumi; KDELC family like 1; KDELCL1; KTEL (Lys-Tyr-Glu-Leu) containing 1; KTEL motif-containing protein 1; KTELC1; MDS010; MDSRP; Myelodysplastic syndromes relative protein; O-glucosyltransferase Rumi homolog; Poglut1; Protein O-glucosyltransferase 1; protein O-xylosyltransferase; Protein O-xylosyltransferase POGLUT1; RGD1306248; Rumi; UNQ490/PRO1006; Wing-shaped neural plate protein; Wsnp; x 010 protein
Gene Symbols POGLUT1
Host Species Rabbit
Immunogen Recombinant Human Protein O-glucosyltransferase 1 protein (132-392AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56983
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.