Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POGZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541563 Supplier Thermo Scientific Supplier No. PA541563
Add to Cart
Add to Cart

POGZ Polyclonal Antibody

POGZ Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen POGZ
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene POGZ
Gene Accession No. Q7Z3K3
Gene Alias 9530006B08Rik; KIAA0461; MRD37; Nbla00003; pogo transposable element derived with ZNF domain; pogo transposable element with ZNF domain; POGZ; putative protein product of Nbla00003; SUHW5; Suppressor of hairy wing homolog 5; WHSUS; Zinc finger protein 280E; zinc finger protein 635; ZNF280E; ZNF635; ZNF635m
Gene Symbols POGZ
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human POGZ (aa 13-62).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23126
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.