Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pogz Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991951
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1951 25 μL
50-199-1950 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1951 Supplier Rockland Immunochemicals Supplier No. 600401A89S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is known to react with mouse Pogz protein. A BLAST analysis was used to suggest cross-reactivity with Pogz from human, dog, short-tailed opossum, cattle, rat, chimpanzee, macaque, olive baboon, and chicken sources based on 100% homology with the immunizing sequence. Reactivity with Pogz from other sources has not been determined.

POGZ appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid systemThe protein encoded by this gene appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid system. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed.

Specifications

Antigen Pogz
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene POGZ
Gene Accession No. Q7Z3K3
Gene Alias 9530006B08Rik; BOS_2943; KIAA0461; MRD37; Nbla00003; pogo transposable element derived with ZNF domain; Pogo transposable element with ZNF domain; POGZ; putative protein product of Nbla00003; SUHW5; Suppressor of hairy wing homolog 5; WHSUS; Zinc finger protein 280E; Zinc finger protein 635; ZNF280E; ZNF635; ZNF635m
Gene Symbols POGZ
Host Species Rabbit
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100856450, 23126, 310658, 429985, 535254, 710634, 747555
Target Species Baboon, Bovine, Canine, Chicken, Chimpanzee, Human, Marsupial, Rat, Rhesus Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.