Learn More
Description
Reagent requires Albumin Standard (23-666-066)/Chemistry Calibrator (23-666-189). HA901-1400 is used for Hitachi 911, 912, 914. HA701-600 is used for Hitachi 704. HA401-3000 is used for Hitachi 747.
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Albumin Standard Reagent Kit |
| For Use With (Application) | Pharmaceuticals |
| Quantity | 15 mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.