Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Albumin Reagents

Catalog No. 23666068 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
15 mL
120 mL
500 mL
1000 mL
Type:
Albumin Reagent Kit
Albumin Standard Reagent Kit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-666-068 120 mL Albumin Reagent Kit
23-666-069 1000 mL Albumin Reagent Kit
23-666-070 500 mL Albumin Reagent Kit
23-666-066 15 mL Albumin Standard Reagent Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-666-068 Supplier Pointe Scientific Supplier No. A7502120
Add to Cart
Add to Cart
This item is not returnable. View return policy

For quantitative determination of Albumin in serum

Reagent requires Albumin Standard (23-666-066)/Chemistry Calibrator (23-666-189). HA901-1400 is used for Hitachi 911, 912, 914. HA701-600 is used for Hitachi 704. HA401-3000 is used for Hitachi 747.

Specifications

Certifications/Compliance CE Marked
Type Albumin Reagent Kit
Formulation Bromocresol green 0.15g/L, Buffer pH 4.6 ±0.1, Surfactants, non-reactive ingredients and Stabilizers, Sodium Azide 0.01%
For Use With (Application) Quantitative determination of Albumin in Serum, in-vitro diagnostic use only
pH 4.66
Quantity 120 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.