Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Alcohol Reagents

Catalog No. 23666072 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
6 x 6.5 mL
10 x 15 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-072 10 x 15 mL
23-666-073 6 x 6.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-666-072 Supplier Pointe Scientific Supplier No. A7504150
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Certifications/Compliance CE Marked
Type Alcohol Reagent Kit
Formulation Alcohol Dehydrogenase (Yeast) 200000U/L, NAD 2.6mM, Buffer pH 9.2 ± 0.1, Surfactant, preservative
For Use With (Application) Quantitative determination of Ethyl Alcohol in Serum or whole Blood, in-vitro diagnostic use only
Quantity 10 x 15 mL
Form Dry Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.